Assay Details
| Gene Symbol: Map1lc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022867.2 | NP_074058.2 | 3 - 4 | 280 | 133 | |
| GenBank mRNA | AY206669.1 | - | 3 - 4 | 242 | 133 | |
| AY310156.1 | - | 808 | 133 | |||
| AY392036.1 | - | 3 - 4 | 280 | 133 | ||
| BC058144.1 | - | 3 - 4 | 238 | 6889616 | 133 | |
| BC083556.1 | - | 3 - 4 | 260 | 7189855 | 133 | |
| FQ221245.1 | - | 3 - 4 | 291 | 133 | ||
| FQ228912.1 | - | 3 - 4 | 291 | 133 | ||
| U05784.1 | - | 3 - 4 | 230 | 133 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Map1lc3b
Genomic Map