Assay Details
| Gene Symbol: Lcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017024.2 | NP_058720.2 | 3 - 4 | 440 | 126 | |
| XM_006255441.1 | XP_006255503.1 | 3 - 4 | 909 | 126 | ||
| GenBank mRNA | BC091155.1 | - | 3 - 4 | 440 | 7370790 | 126 |
| FQ209624.1 | - | 3 - 4 | 455 | 126 | ||
| FQ218245.1 | - | 3 - 4 | 455 | 126 | ||
| FQ218883.1 | - | 3 - 4 | 458 | 126 | ||
| U62803.1 | - | 3 - 4 | 443 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lcat
Genomic Map