Assay Details
| Gene Symbol: Kiaa0895l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044292.1 | NP_001037757.1 | 2 - 3 | 1057 | 60 | |
| XM_008772533.1 | XP_008770755.1 | 2 - 3 | 1715 | 60 | ||
| GenBank mRNA | BC098045.1 | - | 1 - 2 | 628 | 7457085 | 60 |
| BC098942.1 | - | 2 - 3 | 1057 | 7456844 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kiaa0895l
Genomic Map