Assay Details
| Gene Symbol: Kiaa0895l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044292.1 | NP_001037757.1 | 5 - 6 | 1646 | 57 | |
| XM_008772533.1 | XP_008770755.1 | 5 - 6 | 2304 | 57 | ||
| GenBank mRNA | BC098045.1 | - | 4 - 5 | 1217 | 7457085 | 57 |
| BC098942.1 | - | 5 - 6 | 1646 | 7456844 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kiaa0895l
Genomic Map