Assay Details
| Gene Symbol: Kiaa0895l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044292.1 | NP_001037757.1 | 1 - 2 | 440 | 74 | |
| GenBank mRNA | BC098942.1 | - | 1 - 2 | 440 | 7456844 | 74 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC098942.1, NM_001044292.1 transcripts.
Set Membership: |
|

Kiaa0895l
Genomic Map