Assay Details
| Gene Symbol: Prmt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191599.1 | NP_001178528.1 | 3 - 4 | 698 | 157 | |
| XM_006255424.2 | XP_006255486.1 | 3 - 4 | 734 | 157 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prmt10
Genomic Map