Assay Details
| Gene Symbol: Exoc3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106178.1 | NP_001099648.1 | 4 - 5 | 682 | 81 | |
| XM_006255456.2 | XP_006255518.1 | 5 - 6 | 1099 | 81 | ||
| XM_006255457.2 | XP_006255519.1 | 5 - 6 | 1435 | 81 | ||
| XM_006255460.2 | XP_006255522.1 | 4 - 5 | 613 | 81 | ||
| XM_006255461.2 | XP_006255523.1 | 4 - 5 | 562 | 81 | ||
| XM_006255463.1 | XP_006255525.1 | 2 - 3 | 307 | 81 | ||
| XM_008772478.1 | XP_008770700.1 | 5 - 6 | 1318 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Exoc3l1
Genomic Map