Assay Details
| Gene Symbol: Tradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100480.1 | NP_001093950.1 | 2 - 3 | 434 | 70 | |
| XM_006255442.2 | XP_006255504.1 | 3 - 4 | 510 | 70 | ||
| XM_006255444.2 | XP_006255506.1 | 3 - 4 | 509 | 70 | ||
| GenBank mRNA | FQ215613.1 | - | 3 - 4 | 483 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tradd
Genomic Map