Assay Details
| Gene Symbol: Ddx39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053563.2 | NP_446015.2 | 2 - 3 | 512 | 103 | |
| XM_006255314.2 | XP_006255376.1 | 2 - 3 | 412 | 103 | ||
| GenBank mRNA | AF063447.1 | - | 2 - 3 | 308 | 103 | |
| BC086328.1 | - | 2 - 3 | 512 | 7132938 | 103 | |
| FQ231639.1 | - | 2 - 3 | 417 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx39a
Genomic Map