Assay Details
| Gene Symbol: Ddx39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053563.2 | NP_446015.2 | 8 - 9 | 1277 | 119 | |
| XM_006255314.2 | XP_006255376.1 | 8 - 9 | 1177 | 119 | ||
| GenBank mRNA | AF063447.1 | - | 8 - 9 | 1073 | 119 | |
| BC086328.1 | - | 8 - 9 | 1277 | 7132938 | 119 | |
| FQ231639.1 | - | 8 - 9 | 1183 | 119 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ddx39a
Genomic Map