Assay Details
| Gene Symbol: Lrrc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004088.3 | NP_001004088.3 | 4 - 5 | 513 | 83 | |
| XM_006255503.1 | XP_006255565.1 | 4 - 5 | 501 | 83 | ||
| XM_006255504.1 | XP_006255566.1 | 4 - 5 | 501 | 83 | ||
| XM_006255505.1 | XP_006255567.1 | 4 - 5 | 501 | 83 | ||
| XM_006255507.1 | XP_006255569.1 | 4 - 5 | 501 | 83 | ||
| XM_006255508.2 | XP_006255570.1 | 2 - 3 | 666 | 83 | ||
| XM_006255509.2 | XP_006255571.1 | 4 - 5 | 442 | 83 | ||
| GenBank mRNA | AY248698.1 | - | 3 - 4 | 484 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY248698.1, XM_006255508.2 transcripts.
Set Membership: |
|

Lrrc36
Genomic Map