Assay Details
| Gene Symbol: Prkaca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100922.1 | NP_001094392.1 | 5 - 6 | 554 | 148 | |
| XM_006255219.1 | XP_006255281.1 | 5 - 6 | 488 | 148 | ||
| GenBank mRNA | AY375243.1 | - | 5 - 6 | 400 | 148 | |
| BC129128.1 | - | 5 - 6 | 554 | 7314651 | 148 | |
| X57986.1 | - | 5 - 6 | 464 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkaca
Genomic Map