Assay Details
| Gene Symbol: Gas8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039030.1 | NP_001034119.1 | 6 - 7 | 784 | 65 | |
| XM_006255777.2 | XP_006255839.1 | 6 - 7 | 825 | 65 | ||
| XM_006255779.2 | XP_006255841.1 | 6 - 7 | 1366 | 65 | ||
| XM_008772635.1 | XP_008770857.1 | 3 - 4 | 416 | 65 | ||
| XM_008772636.1 | XP_008770858.1 | 3 - 4 | 352 | 65 | ||
| XM_008772637.1 | XP_008770859.1 | 3 - 4 | 344 | 65 | ||
| GenBank mRNA | BC099759.1 | - | 6 - 7 | 784 | 7929646 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Gas8
Genomic Map