Assay Details
| Gene Symbol: Pabpn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113779.1 | NP_001107251.1 | 5 - 6 | 667 | 68 | |
| GenBank mRNA | BC158789.1 | - | 5 - 6 | 667 | 7306904 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Pabpn1l
Genomic Map