Assay Details
| Gene Symbol: LOC100909612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751860.3 | XP_003751908.2 | 1 - 2 | 256 | 81 | |
| Gene Symbol: Ucp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012682.2 | NP_036814.1 | 3 - 4 | 705 | 81 | |
| GenBank mRNA | BC088156.1 | - | 3 - 4 | 693 | 7380759 | 81 |
| M11814.1 | - | 3 - 4 | 703 | 81 | ||
| X03894.1 | - | 3 - 4 | 615 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100909612
Genomic Map