Assay Details
| Gene Symbol: Abce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108446.1 | NP_001101916.1 | 2 - 3 | 233 | 144 | |
| XM_006255407.2 | XP_006255469.1 | 2 - 3 | 250 | 144 | ||
| GenBank mRNA | AY724536.1 | - | 2 - 3 | 233 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abce1
Genomic Map