Assay Details
| Gene Symbol: Cbfa2t3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108453.1 | NP_001101923.1 | 8 - 9 | 1225 | 73 | |
| XM_006255770.2 | XP_006255832.1 | 7 - 8 | 2119 | 73 | ||
| XM_006255772.2 | XP_006255834.1 | 8 - 9 | 1380 | 73 | ||
| XM_008772629.1 | XP_008770851.1 | 7 - 8 | 3090 | 73 | ||
| XM_008772630.1 | XP_008770852.1 | 6 - 7 | 3013 | 73 | ||
| XM_008772631.1 | XP_008770853.1 | 7 - 8 | 1575 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cbfa2t3
Genomic Map