Assay Details
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106670.1 | NP_001100140.1 | 3 - 4 | 353 | 67 | |
GenBank mRNA | FQ211202.1 | - | 3 - 4 | 432 | 67 | |
FQ211244.1 | - | 3 - 4 | 432 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
