Assay Details
| Gene Symbol: Tm2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108670.1 | NP_001102140.1 | 7 - 8 | 1065 | 115 | |
| XM_006238417.2 | XP_006238479.1 | 7 - 8 | 947 | 115 | ||
| XM_006238418.2 | XP_006238480.1 | 7 - 8 | 947 | 115 | ||
| XM_006238419.2 | XP_006238481.1 | 6 - 7 | 842 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tm2d1
Genomic Map