Assay Details
Gene Symbol: RGD1564074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763964.1 | XP_008762186.1 | 14 - 15 | 1838 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
