Assay Details
| Gene Symbol: Elavl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238385.2 | XP_006238447.1 | 7 - 8 | 1044 | 139 | |
| XM_006238388.2 | XP_006238450.1 | 6 - 7 | 847 | 139 | ||
| XM_006238389.2 | XP_006238451.1 | 6 - 7 | 1384 | 139 | ||
| XM_006238391.2 | 6 - 7 | 1384 | 136 | |||
| XM_006238395.2 | XP_006238457.1 | 5 - 6 | 775 | 139 | ||
| XM_008763848.1 | XP_008762070.1 | 5 - 6 | 1090 | 139 | ||
| XM_008763849.1 | XP_008762071.1 | 5 - 6 | 1090 | 136 | ||
| XM_008763850.1 | XP_008762072.1 | 5 - 6 | 753 | 139 | ||
| GenBank mRNA | AB098713.1 | - | 6 - 7 | 1308 | 136 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P841017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121397 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P841017 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P841017 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P841017 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P143503 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P841017 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61373 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44994 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63843 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28585 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33300 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19078 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82658 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81959 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50838 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85811 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10878023 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878020 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878025 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878019 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878021 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878017 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878022 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878018 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10878016 | - | - |
| Rat Genome 230 2.0 Array | 1383736_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383736_at | - | - |
| Rat Genome 230 2.0 Array | 1383768_at | - | - |
| Rat Genome 230 2.0 Array | 1383768_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381677_at | - | - |
| Rat Genome 230 2.0 Array | 1381677_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383768_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383768_at | - | - |
| Affymetrix HT Rat Focus Array | 1383736_at | - | - |
| Affymetrix HT Rat Focus Array | 1383736_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381677_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381677_at | - | - |
| Rat Expression Set 230 | 1383736_at | - | - |
| Rat Expression Set 230 | 1383768_PM_at | - | - |
| Rat Expression Set 230 | 1383768_at | - | - |
| Rat Expression Set 230 | 1383736_PM_at | - | - |
| Rat Expression Set 230 | 1381677_at | - | - |
| Rat Expression Set 230 | 1381677_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878016 | - | - |
| Rat Genome U34 Set | rc_AA997244_at | - | - |

Elavl2
Genomic Map