Assay Details
| Gene Symbol: Ascc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007632.2 | NP_001007633.1 | 9 - 10 | 1021 | 105 | |
| XM_006256404.2 | XP_006256466.1 | 9 - 10 | 1215 | 105 | ||
| XM_006256405.2 | XP_006256467.1 | 9 - 10 | 1212 | 105 | ||
| XM_006256407.2 | XP_006256469.2 | 9 - 10 | 1101 | 105 | ||
| XM_006256408.2 | XP_006256470.1 | 9 - 10 | 1170 | 105 | ||
| XM_006256409.1 | XP_006256471.1 | 8 - 9 | 956 | 105 | ||
| XM_006256410.2 | XP_006256472.1 | 9 - 10 | 1167 | 105 | ||
| XM_008772881.1 | XP_008771103.1 | 8 - 9 | 953 | 105 | ||
| GenBank mRNA | BP504504.1 | - | 5 - 6 | 512 | 105 | |
| FQ226270.1 | - | 6 - 7 | 668 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P055843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9268 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055843 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055843 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P055843 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P279095 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36250 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79708 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45250 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55717 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60419 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10829958 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829955 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829962 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829959 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829956 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829964 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829963 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829961 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829960 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829957 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10829954 | - | - |
| Rat Genome 230 2.0 Array | 1375907_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375907_at | - | - |
| Affymetrix HT Rat Focus Array | 1375907_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375907_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000005779.rn.1 | TC2000000655.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375907_PM_at | - | - |
| Rat Expression Set 230 | 1375907_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829954 | - | - |
| Rat Genome U34 Set | rc_AI044617_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21408363 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349263 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62660 | - | - |

Ascc1
Genomic Map