Assay Details
| Gene Symbol: Trappc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728958.3 | XP_002729004.1 | 4 - 5 | 574 | 92 | |
| XM_008772952.1 | XP_008771174.1 | 5 - 6 | 511 | 92 | ||
| XM_008772953.1 | XP_008771175.1 | 4 - 5 | 423 | 92 | ||
More Information
Set Membership: |
|

Trappc3l
Genomic Map