Assay Details
| Gene Symbol: Npffr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022291.2 | NP_071627.2 | 1 - 2 | 333 | 76 | |
| XM_006256485.2 | XP_006256547.1 | 1 - 2 | 479 | 76 | ||
| GenBank mRNA | AB040103.1 | - | 1 - 2 | 325 | 76 | |
| AF268901.1 | - | 1 - 2 | 325 | 76 | ||
| AF330056.1 | - | 1 - 2 | 325 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npffr1
Genomic Map