Assay Details
| Gene Symbol: LOC100911315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772970.1 | XP_008771192.1 | 3 - 4 | 477 | 71 | |
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106388.1 | NP_001099858.1 | 11 - 12 | 1242 | 71 | |
| XM_006256444.2 | XP_006256506.1 | 14 - 15 | 1544 | 71 | ||
| XM_006256445.2 | XP_006256507.1 | 7 - 8 | 795 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911315
Genomic Map