Assay Details
| Gene Symbol: LOC100911315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772970.1 | XP_008771192.1 | 1 - 2 | 254 | 80 | |
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106388.1 | NP_001099858.1 | 9 - 10 | 1019 | 80 | |
| XM_006256444.2 | XP_006256506.1 | 12 - 13 | 1321 | 80 | ||
| XM_006256445.2 | XP_006256507.1 | 5 - 6 | 572 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911315
Genomic Map