Assay Details
| Gene Symbol: Ddx50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013198.2 | NP_001013216.2 | 14 - 15 | 2032 | 276 | |
| GenBank mRNA | BC091427.1 | - | 8 - 9 | 918 | 7366491 | 276 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P344863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104830 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P344863 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P344863 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P344863 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P344863 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34432 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85025 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10833229 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833231 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833219 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833218 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833226 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833228 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833230 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833217 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833225 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833227 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833222 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833223 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833221 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833220 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833216 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833224 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10833215 | - | - |
| Rat Genome 230 2.0 Array | 1390688_at | - | - |
| Rat Genome 230 2.0 Array | 1381598_at | - | - |
| Rat Genome 230 2.0 Array | 1390688_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391395_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391395_at | - | - |
| Rat Genome 230 2.0 Array | 1381598_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381598_at | - | - |
| Affymetrix HT Rat Focus Array | 1390688_at | - | - |
| Affymetrix HT Rat Focus Array | 1391395_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390688_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381598_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391395_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000012844.rn.1 | TC2000001566.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012842.rn.1 | TC2000001566.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000012845.rn.1 | TC2000001566.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390688_PM_at | - | - |
| Rat Expression Set 230 | 1390688_at | - | - |
| Rat Expression Set 230 | 1381598_PM_at | - | - |
| Rat Expression Set 230 | 1381598_at | - | - |
| Rat Expression Set 230 | 1391395_PM_at | - | - |
| Rat Expression Set 230 | 1391395_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10833215 | - | - |
| Rat Genome U34 Set | rc_AA799576_at | - | - |
| Rat Genome U34 Set | rc_AA818165_at | - | - |
| Rat Genome U34 Set | rc_AI170715_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22392510 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355152 | - | - |

Ddx50
Genomic Map