Assay Details
| Gene Symbol: Ddx50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013198.2 | NP_001013216.2 | 8 - 9 | 1337 | 142 | |
| GenBank mRNA | BC091427.1 | - | 2 - 3 | 223 | 7366491 | 142 |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx50
Genomic Map