Assay Details
| Gene Symbol: Spock2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108533.2 | NP_001102003.1 | 4 - 5 | 364 | 63 | |
| XM_006256419.2 | XP_006256481.1 | 4 - 5 | 1005 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spock2
Genomic Map