Assay Details
Gene Symbol: Lrrc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109171.1 | NP_001102641.1 | 4 - 5 | 538 | 121 | |
XM_008772902.1 | XP_008771124.1 | 3 - 4 | 360 | 121 | ||
XM_008772903.1 | XP_008771125.1 | 3 - 4 | 1327 | 121 | ||
GenBank mRNA | FQ223562.1 | - | 4 - 5 | 532 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P042639 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P649046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100536 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042639 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P649046 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P649046 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042639 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P649046 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P042639 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68311 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10830033 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830032 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830031 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10830030 | - | - |
Rat Genome 230 2.0 Array | 1373167_at | - | - |
Rat Genome 230 2.0 Array | 1373167_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373167_at | - | - |
Affymetrix HT Rat Focus Array | 1373167_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2000005983.rn.1 | TC2000000682.rn.1 | RTA TC |
Rat Expression Set 230 | 1373167_at | - | - |
Rat Expression Set 230 | 1373167_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830030 | - | - |
Rat Genome U34 Set | rc_AI171557_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21875503 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370809 | - | - |
