Assay Details
| Gene Symbol: Ccdc104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024866.1 | NP_001020037.1 | 3 - 4 | 400 | 60 | |
| GenBank mRNA | BC097425.1 | - | 3 - 4 | 400 | 7455112 | 60 |
| FQ213878.1 | - | 3 - 4 | 237 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc104
Genomic Map