Assay Details
| Gene Symbol: Ccdc104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024866.1 | NP_001020037.1 | 4 - 5 | 519 | 72 | |
| XM_006251598.2 | XP_006251660.1 | 3 - 4 | 511 | 72 | ||
| GenBank mRNA | BC097425.1 | - | 4 - 5 | 519 | 7455112 | 72 |
| FQ213878.1 | - | 4 - 5 | 356 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc104
Genomic Map