Assay Details
| Gene Symbol: Ccdc104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024866.1 | NP_001020037.1 | 6 - 7 | 652 | 68 | |
| XM_006251598.2 | XP_006251660.1 | 5 - 6 | 644 | 68 | ||
| GenBank mRNA | BC097425.1 | - | 6 - 7 | 652 | 7455112 | 68 |
| FQ213878.1 | - | 6 - 7 | 489 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc104
Genomic Map