Assay Details
| Gene Symbol: Rab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031090.2 | NP_112352.2 | 1 - 2 | 190 | 71 | |
| GenBank mRNA | BC066662.1 | - | 1 - 2 | 190 | 5623442 | 71 |
| FQ209441.1 | - | 1 - 2 | 200 | 71 | ||
| FQ213656.1 | - | 1 - 2 | 200 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC066662.1,NM_031090.2 transcripts.
This assay is in the 5'UTR region of the BC066662.1,NM_031090.2 transcripts.
Set Membership: |
|

Rab1a
Genomic Map