Assay Details
| Gene Symbol: Rab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031090.2 | NP_112352.2 | 3 - 4 | 376 | 130 | |
| GenBank mRNA | AY321327.1 | - | 332 | 130 | ||
| BC066662.1 | - | 3 - 4 | 376 | 5623442 | 130 | |
| FQ209441.1 | - | 3 - 4 | 386 | 130 | ||
| FQ213656.1 | - | 3 - 4 | 386 | 130 | ||
| FQ214472.1 | - | 3 - 4 | 386 | 130 | ||
| J02998.1 | - | 3 - 4 | 199 | 130 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rab1a
Genomic Map