Assay Details
| Gene Symbol: Rab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031090.2 | NP_112352.2 | 4 - 5 | 465 | 116 | |
| GenBank mRNA | AY321327.1 | - | 421 | 116 | ||
| BC066662.1 | - | 4 - 5 | 465 | 5623442 | 116 | |
| FQ209441.1 | - | 4 - 5 | 475 | 116 | ||
| FQ213656.1 | - | 4 - 5 | 475 | 116 | ||
| FQ214472.1 | - | 4 - 5 | 475 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rab1a
Genomic Map