Assay Details
| Gene Symbol: Lgalsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134730.1 | NP_001128202.1 | 5 | 107 | ||
| GenBank mRNA | BC168193.1 | - | 5 | 7109522 | 107 | |
| FQ211679.1 | - | 5 | 107 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lgalsl
Genomic Map