Assay Details
| Gene Symbol: Prl8a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134385.2 | NP_599212.1 | 3 - 4 | 283 | 130 | |
| XM_006253904.1 | XP_006253966.1 | 3 - 4 | 274 | 130 | ||
| GenBank mRNA | AF239745.1 | - | 3 - 4 | 283 | 130 | |
| AF239746.1 | - | 3 - 4 | 280 | 130 | ||
| AF239747.1 | - | 3 - 4 | 283 | 130 | ||
| BC100138.1 | - | 3 - 4 | 276 | 7442617 | 130 | |
| BC127474.1 | - | 3 - 4 | 274 | 7443385 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prl8a9
Genomic Map