Assay Details
| Gene Symbol: Eef1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106106.1 | NP_001099576.1 | 1 - 2 | 110 | 131 | |
| GenBank mRNA | BC166841.1 | - | 98 | 9027959 | 131 | |
| FQ212823.1 | - | 1 - 2 | 97 | 131 | ||
| FQ219292.1 | - | 1 - 2 | 129 | 131 | ||
| FQ223579.1 | - | 1 - 2 | 97 | 131 | ||
| FQ225943.1 | - | 1 - 2 | 129 | 131 | ||
| FQ229476.1 | - | 1 - 2 | 129 | 131 | ||
| FQ231336.1 | - | 1 - 2 | 97 | 131 | ||
| FQ233549.1 | - | 1 - 2 | 97 | 131 | ||
| FQ234239.1 | - | 1 - 2 | 109 | 131 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eef1e1
Genomic Map