Assay Details
| Gene Symbol: Prl2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138861.1 | NP_620216.1 | 2 - 3 | 267 | 151 | |
| GenBank mRNA | AB022882.1 | - | 2 - 3 | 264 | 151 | |
| AF234635.1 | - | 2 - 3 | 267 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prl2b1
Genomic Map