Assay Details
| Gene Symbol: Txndc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271330.1 | NP_001258259.1 | 9 - 10 | 1205 | 93 | |
| GenBank mRNA | FM093163.1 | - | 2 - 3 | 163 | 93 | |
More Information
Set Membership: |
|

Txndc5
Genomic Map