Assay Details
| Gene Symbol: Ptpdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106104.1 | NP_001099574.1 | 3 - 4 | 522 | 60 | |
| XM_006253700.2 | XP_006253762.1 | 2 - 3 | 1345 | 60 | ||
| XM_006253701.2 | XP_006253763.1 | 3 - 4 | 1042 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpdc1
Genomic Map