Assay Details
| Gene Symbol: Gfod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170334.1 | NP_001163805.1 | 1 - 2 | 251 | 68 | |
| XM_006253805.2 | XP_006253867.1 | 1 - 2 | 940 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gfod1
Genomic Map