Assay Details
| Gene Symbol: Ly86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106128.1 | NP_001099598.1 | 2 - 3 | 322 | 147 | |
| XM_006253800.2 | XP_006253862.1 | 3 - 4 | 494 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ly86
Genomic Map