Assay Details
| Gene Symbol: Ly86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106128.1 | NP_001099598.1 | 4 - 5 | 510 | 80 | |
| XM_006253800.2 | XP_006253862.1 | 5 - 6 | 682 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P123622 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P130898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121579 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P123622 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P130898 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P130898 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P123622 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P130898 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P123622 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P130898 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13913 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70353 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10798032 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798028 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798031 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798029 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798030 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10798027 | - | - |
| Rat Genome 230 2.0 Array | 1393280_at | - | - |
| Rat Genome 230 2.0 Array | 1393280_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393280_at | - | - |
| Affymetrix HT Rat Focus Array | 1393280_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700011203.rn.1 | TC1700001617.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700011204.rn.1 | TC1700001617.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393280_PM_at | - | - |
| Rat Expression Set 230 | 1393280_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798027 | - | - |
| Rat Genome U34 Set | rc_AA874924_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20789084 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358738 | - | - |

Ly86
Genomic Map