Assay Details
| Gene Symbol: Prl8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022846.2 | NP_074037.1 | 3 - 4 | 252 | 163 | |
| XM_006253906.1 | XP_006253968.1 | 3 - 4 | 312 | 163 | ||
| GenBank mRNA | BC097316.1 | - | 3 - 4 | 253 | 7442457 | 163 |
| FQ219955.1 | - | 3 - 4 | 312 | 163 | ||
| FQ220067.1 | - | 3 - 4 | 311 | 163 | ||
| FQ220110.1 | - | 3 - 4 | 312 | 163 | ||
| FQ229664.1 | - | 3 - 4 | 163 | |||
| FQ229702.1 | - | 3 - 4 | 310 | 163 | ||
| FQ229765.1 | - | 3 - 4 | 309 | 163 | ||
| FQ229882.1 | - | 3 - 4 | 163 | |||
| L06441.1 | - | 3 - 4 | 254 | 163 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prl8a2
Genomic Map