Assay Details
Gene Symbol: LOC103689971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253876.2 | XP_006253938.1 | 1776 | 154 | ||
XM_006253878.2 | XP_006253940.1 | 1659 | 154 | |||
XM_006253880.2 | XP_006253942.1 | 1587 | 154 | |||
XM_006253881.2 | XP_006253943.1 | 1650 | 154 | |||
XM_008771607.1 | XP_008769829.1 | 1704 | 154 |
Gene Symbol: Exoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134414.2 | NP_599241.2 | 14 - 15 | 1704 | 154 | |
XM_006253891.1 | XP_006253953.1 | 15 - 16 | 1776 | 154 | ||
XM_006253893.2 | XP_006253955.1 | 14 - 15 | 1660 | 154 | ||
XM_006253895.2 | XP_006253957.1 | 13 - 14 | 1588 | 154 | ||
GenBank mRNA | AF032666.1 | - | 14 - 15 | 1642 | 154 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
