Assay Details
| Gene Symbol: Serpinb6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012214.1 | NP_001012214.1 | 2 - 3 | 192 | 61 | |
| GenBank mRNA | BC079108.1 | - | 2 - 3 | 192 | 7125468 | 61 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Serpinb6b
Genomic Map