Assay Details
| Gene Symbol: Serpinb6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012214.1 | NP_001012214.1 | 5 - 6 | 615 | 131 | |
| GenBank mRNA | BC079108.1 | - | 5 - 6 | 615 | 7125468 | 131 |
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb6b
Genomic Map