Assay Details
| Gene Symbol: Prl7b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153738.1 | NP_714960.1 | 3 - 4 | 287 | 121 | |
| GenBank mRNA | AF525159.1 | - | 3 - 4 | 287 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prl7b1
Genomic Map